missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Alfa Aesar™

Brand:  Alfa Aesar 030067.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11392738

  • £125.08 / 1000mm


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
1000mm
One end closed
Videos
SDS