missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Alfa Aesar™

Brand:  Alfa Aesar 030067.EM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 800mm

 View more versions of this product

Product Code. 11332748

  • £99.64 / 800mm


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
800mm
One end closed
Videos
SDS