missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 030067.EH

Al-23 Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11382738

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.