missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Alfa Aesar™

Brand:  Alfa Aesar 030067.DR

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 200mm

 View more versions of this product

Product Code. 11372738

  • £60.53 / 200mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
200mm
One end closed
Videos
SDS