missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 80mm, I.D.: 70mm, Alfa Aesar™

Brand:  Alfa Aesar 030089.DM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11382848

  • £633.46 / 100mm


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Specifications

Al-23 Tube
100mm
One end closed
Videos
SDS