missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 40mm, I.D.: 34mm, Alfa Aesar™

Brand:  Alfa Aesar 030082.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11322818

  • £266.48 / 300mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
300mm
One end closed
Videos
SDS