missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 25mm, I.D.: 20mm, Alfa Aesar™

Brand:  Alfa Aesar 030079.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11392798

  • £137.80 / 100mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
100mm
One end closed
Videos
SDS