missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube for Heating Coils, O.D.: 38mm, I.D.: 29mm, Pitch: 2.0, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 033207.DR

Al-23 Tube for Heating Coils, O.D.: 38mm, I.D.: 29mm, Pitch: 2.0, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11323158

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.