missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube for Heating Coils, O.D.: 21mm, I.D.: 15mm, Pitch: 1.5, Alfa Aesar™

Brand:  Alfa Aesar 033204.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11353138

  • £132.50 / 100mm


Estimated Shipment date
from Supplier 07-11-2017
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube for Heating Coils

Specifications

Al-23 Tube for Heating Coils
100mm
Documentation

Technical Documents