missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube for Heating Coils, O.D.: 21mm, I.D.: 15mm, Pitch: 1.5, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 033204.DW

Al-23 Tube for Heating Coils, O.D.: 21mm, I.D.: 15mm, Pitch: 1.5, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11373138

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.