missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open (Thin walls), O.D.: 6mm, I.D.: 5mm, Alfa Aesar™

Brand:  Alfa Aesar 031706.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11318247

  • £21.40 / 100mm


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open (Thin walls)

Specifications

Al-23 Tube
100mm
Both ends open (Thin walls)
Videos
SDS