missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open (Thin walls), O.D.: 4mm, I.D.: 3mm, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 031704.DW

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11324177

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Tube
Both ends open (Thin walls)
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers