missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open (Thin walls), O.D.: 4mm, I.D.: 3mm, Alfa Aesar™

Brand:  Alfa Aesar 031704.DR

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 200mm

 View more versions of this product

Product Code. 11304177

  • £28.30 / 200mm


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open (Thin walls)

Specifications

Al-23 Tube
200mm
Both ends open (Thin walls)
Videos
SDS