missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open (Thin walls), O.D.: 16mm, I.D.: 15mm, Alfa Aesar™

Brand:  Alfa Aesar 031711.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11308267

  • £33.50 / 100mm


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open (Thin walls)

Specifications

Al-23 Tube
100mm
Both ends open (Thin walls)
Videos
SDS