missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open, O.D.: 70mm, I.D.: 60mm, Alfa Aesar™

Brand:  Alfa Aesar 031584.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11334067

  • £129.53 / 100mm


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
100mm
Both ends open
Videos
SDS