missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 6mm, I.D.: 3mm, Alfa Aesar™

Brand:  Alfa Aesar 031556.EM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 800mm

 View more versions of this product

Product Code. 11313937

  • £97.70 / 800mm


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
800mm
Both ends open
Videos
SDS