missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both ends open, O.D.: 65mm, I.D.: 55mm, Alfa Aesar™

Brand:  Alfa Aesar 031582.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11354057

  • £1056.71 / 1000mm


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
1000mm
Both ends open
Videos
SDS