missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 45mm, I.D.: 38mm, Alfa Aesar™

Brand:  Alfa Aesar 031578.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11344027

  • £163.56 / 300mm


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
300mm
Both ends open
Videos
SDS