missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 20mm, I.D.: 15mm, Alfa Aesar™

Brand:  Alfa Aesar 031570.EH

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 600mm

 View more versions of this product

Product Code. 11363987

  • £175.85 / 600mm


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
600mm
Both ends open
Videos
SDS