missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Alfa Aesar 031566.ED

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 500mm

 View more versions of this product

Product Code. 15421207

  • £76.74 / 500mm


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
500mm
Both ends open
Videos
SDS