missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Solid Rod, Round, Dia.: 8mm, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 032935.EO

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11323098

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Solid Rod
1000 mm
Rod
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers