missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Solid Rod, Round, Dia.: 5mm, Thermo Scientific Chemicals
POA - £131.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363078
|
Thermo Scientific Alfa Aesar
032931.DR |
200 mm |
£43.60
200mm |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11353078
|
Thermo Scientific Alfa Aesar
032931.DT |
250 mm |
£50.40
250mm |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11333078
|
Thermo Scientific Alfa Aesar
032931.DW |
300 mm |
£56.20
300mm |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11303088
|
Thermo Scientific Alfa Aesar
032931.EH |
600 mm |
£102.00
600mm |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15421297
|
Thermo Scientific Alfa Aesar
032931.Q@ |
400 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11343078
|
Thermo Scientific Alfa Aesar
032931.DM |
100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11383078
|
Thermo Scientific Alfa Aesar
032931.EM |
800 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11393078
|
Thermo Scientific Alfa Aesar
032931.EO |
1000 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only