missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Solid Rod, Round, Dia.: 12mm, Alfa Aesar™

Brand:  Alfa Aesar 032939.EH

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 600mm

 View more versions of this product

Product Code. 11363108

  • £69.11 / 600mm


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Solid Rod
Solid Round Rod

Specifications

Al-23 Solid Rod
600mm
Solid Round Rod
Videos
SDS