missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Wall.: 3mm, Alfa Aesar™

Brand:  Alfa Aesar 030054.FA

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 2030mm

 View more versions of this product

Product Code. 11382708

  • £598.48 / 2030mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
2030mm
One end closed
Videos
SDS