missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Wall.: 3mm, Alfa Aesar™

Brand:  Alfa Aesar 030054.DU

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 270mm

 View more versions of this product

Product Code. 11362708

  • £110.66 / 270mm


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
270mm
One end closed
Videos
SDS