missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 20mm, I.D.: 15mm, Wall.: 2.5mm, Alfa Aesar™

Brand:  Alfa Aesar 030052.EV

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1430mm

 View more versions of this product

Product Code. 11302708

  • £348.60 / 1430mm


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
1430mm
One end closed
Videos
SDS