missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Wall.: 2.5mm, Alfa Aesar™

Brand:  Alfa Aesar 030050.FA

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 2030mm

 View more versions of this product

Product Code. 11302698

  • £284.50 / 2030mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
2030mm
One end closed
Videos
SDS