missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Wall.: 2.5mm, Alfa Aesar™

Brand:  Alfa Aesar 030050.DY

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 375mm

 View more versions of this product

Product Code. 11352688

  • £54.38 / 375mm


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
375mm
One end closed
Videos
SDS