missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 12mm, I.D.: 8mm, Wall.: 2mm, Alfa Aesar™

Brand:  Alfa Aesar 030048.EW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1460mm

 View more versions of this product

Product Code. 15471137

  • £276.40 / 1460mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Protective Tube
One end closed

Specifications

Al-23 Protective Tube
1460mm
One end closed
Videos
SDS