missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Oval Cross Section 2-Bore Insulators, O.D.: 2.3mm/1.4mm, Two 0.7mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032777.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11353008

  • £14.20 / 100mm


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Oval Cross Section 2-Bore Insulator
Two bores

Specifications

Al-23 Oval Cross Section 2-Bore Insulator
100mm
Two bores
Videos
SDS