missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Oval Cross Section 2-Bore Insulators, O.D.: 3.0mm/1.5mm, Two 0.8mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032774.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11342998

  • £31.48 / 300mm


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Oval Cross Section 2-Bore Insulator
Two bores

Specifications

Al-23 Oval Cross Section 2-Bore Insulator
300mm
Two bores
Videos
SDS