missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 8mm, Two 1.6mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032658.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11392928

  • £90.95 / 1000mm


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Two bores

Specifications

Al-23 Multibore Round Tube
1000mm
Two bores
Videos
SDS