missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Multibore Round Tube, O.D.: 6mm, Two 1.5mm Bores, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 032656.DM

Al-23 Multibore Round Tube, O.D.: 6mm, Two 1.5mm Bores, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11362928

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.