missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 6mm, Two 1.5mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032656.DT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 250mm

 View more versions of this product

Product Code. 11322928

  • £48.34 / 250mm


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Two bores

Specifications

Al-23 Multibore Round Tube
250mm
Two bores
Videos
SDS