Learn More
Al-23 Multibore Round Tube, O.D.: 5mm, Two 1.2mm Bores, Thermo Scientific™
POA - £57.66
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11332918
|
Thermo Scientific Alfa Aesar
032654.DM |
100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11322918
|
Thermo Scientific Alfa Aesar
032654.DR |
200 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11342918
|
Thermo Scientific Alfa Aesar
032654.DT |
250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11312918
|
Thermo Scientific Alfa Aesar
032654.DW |
300 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11382918
|
Thermo Scientific Alfa Aesar
032654.EH |
600 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11362918
|
Thermo Scientific Alfa Aesar
032654.EM |
800 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11352918
|
Thermo Scientific Alfa Aesar
032654.EO |
1000 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||