missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 5mm, Two 1.2mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032654.DW

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11312918

  • £21.10 / 300mm


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Specifications

Al-23 Multibore Round Tube
300mm
Two bores
Videos
SDS