missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 2.35mm, Four 0.5mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032602.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11348517

  • £32.75 / 300mm


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Four bores

Specifications

Al-23 Multibore Round Tube
300mm
Four bores
Videos
SDS