missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 2.35mm, Four 0.5mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032602.DR

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 200mm

 View more versions of this product

Product Code. 11338517

  • £22.58 / 200mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Four bores

Specifications

Al-23 Multibore Round Tube
200mm
Four bores
Videos
SDS