missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 2.35mm, Four 0.5mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032602.DT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 250mm

 View more versions of this product

Product Code. 11328517

  • £26.82 / 250mm


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Four bores

Specifications

Al-23 Multibore Round Tube
250mm
Four bores
Videos
SDS