missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 1.2mm, Two 0.3mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032565.DR

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 200mm

 View more versions of this product

Product Code. 11308477

  • £28.00 / 200mm


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Two bores

Specifications

Al-23 Multibore Round Tube
200mm
Two bores
Videos
SDS