missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 1.2mm, Four 0.2mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032600.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11368507

  • £21.84 / 300mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Four bores

Specifications

Al-23 Multibore Round Tube
300mm
Four bores
Videos
SDS