missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Multibore Round Tube, O.D.: 1.2mm, Four 0.2mm Bores, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 032600.DR

Al-23 Multibore Round Tube, O.D.: 1.2mm, Four 0.2mm Bores, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11338507

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.