missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Insulating Tube, O. D.: 4mm, Two 0.8mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032550.EW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1460mm

 View more versions of this product

Product Code. 11308417

  • £118.10 / 1460mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Insulating Tube
Two bores

Specifications

Al-23 Multibore Insulating Tube
1460mm
Two bores
Videos
SDS