missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Grinding Pieces, H: 30mm, 15 Pieces/Kg, Alfa Aesar™

Brand:  Alfa Aesar 033193.A1

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11353128

  • £61.59 / 1kg


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Specifications

Al-23 Grinding Pieces
1kg
Grinding Pieces
Videos
SDS