missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Grinding Pieces, H: 20mm, 53 Pieces/Kg, Alfa Aesar™
Brand: Alfa Aesar 033192.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity | 1kg |
---|
Chemical Identifiers
Al-23 Grinding Pieces | |
Grinding Pieces |
Specifications
Al-23 Grinding Pieces | |
1kg | |
Grinding Pieces |