missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Grinding Pieces, H: 15mm, 157 Pieces/Kg, Alfa Aesar™

Brand:  Alfa Aesar 033191.A1

Code : NEW

Additional Details : Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15441307

  • £240.09 / 1kg


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Grinding Pieces
Grinding Pieces

Specifications

Al-23 Grinding Pieces
1kg
Grinding Pieces
Videos
SDS