missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1A1 Antibody (OTI6E3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71591MFV500
This item is not returnable.
View return policy
Description
AKR1A1 Monoclonal antibody specifically detects AKR1A1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| AKR1A1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI6E3 | |
| Flow Cytometry | |
| alcohol dehydrogenase, alcohol dehydrogenase [NADP+], Aldehyde reductase, Aldo-keto reductase family 1 member A1, aldo-keto reductase family 1, member A1 (aldehyde reductase), ALDR1, ALRMGC1380, ARM, DD3, dihydrodiol dehydrogenase 3, EC 1.1.1, EC 1.1.1.2, MGC12529 | |
| Full length human recombinant protein of human AKR1A1 (NP_006057) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 10327 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction