missing translation for 'onlineSavingsMsg'
Learn More

(+)-AJ 76 hydrochloride, Tocris Bioscience™

Antagonist; preferential action at D2-like autoreceptors

Brand:  Tocris (Bio-Techne) 067810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467913

  • £102.90 / 10mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dopamine receptor antagonist with preferential action at presynaptic receptors (pKi values are 6.95, 6.67, 6.37, 6.21 and 6.07 at hD3. hD4, hD2S, hD2L and rD2 receptors respectively).
Specifications

Chemical Identifiers

85378-82-1
269.813
+-aj 76 hydrochloride, 1s,2r-5-methoxy-1-methyl-n-propyl-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride, cis-+-5-methoxy-1-methyl-2-propylamino tetralin hydrochloride, 1s,2r-5-methoxy-1-methyl-2-propylamino tetralin hydrochloride, 1s,2r-5-methoxy-1-methyl-n-propyl-1,2,3,4-tetrahydronaphthalen-2-aminium chloride, 1s,2r-cis-5-methoxy-1-methyl-2-n-propylamino tetralin hydrochloride
CHEBI:64115
CCCNC1CCC2=C(C1C)C=CC=C2OC.Cl
C15H24ClNO
KIRYNZFMOLYYQB-YECZQDJWSA-N
13755572
(1S,2R)-5-methoxy-1-methyl-N-propyl-1,2,3,4-tetrahydronaphthalen-2-amine;hydrochloride

Specifications

10mg
85378-82-1
+-aj 76 hydrochloride, 1s,2r-5-methoxy-1-methyl-n-propyl-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride, cis-+-5-methoxy-1-methyl-2-propylamino tetralin hydrochloride, 1s,2r-5-methoxy-1-methyl-2-propylamino tetralin hydrochloride, 1s,2r-5-methoxy-1-methyl-n-propyl-1,2,3,4-tetrahydronaphthalen-2-aminium chloride, 1s,2r-cis-5-methoxy-1-methyl-2-n-propylamino tetralin hydrochloride
CCCNC1CCC2=C(C1C)C=CC=C2OC.Cl
269.813
CHEBI:64115
>98%
(+)-AJ 76 hydrochloride
C15H24ClNO
KIRYNZFMOLYYQB-YECZQDJWSA-N
(1S,2R)-5-methoxy-1-methyl-N-propyl-1,2,3,4-tetrahydronaphthalen-2-amine;hydrochloride
13755572
269.81
Videos
Documents