missing translation for 'onlineSavingsMsg'
Learn More

AIRE Rat anti-Human, Clone: TM-724, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9534-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15597596

  • £104.00 / 25µg

Estimated Shipment
05-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. AIRE is a transcriptional regulator protein that binds to DNA as a dimer and tetramer, but not as a monomer, and may be involved in the regulation of gene expression. A nuclear protein, AIRE is a 545 amino acid protein with an HSR domain, two PHD type zinc fingers, and a SAND domain, and may be present in soluble high-molecular-weight complexes. It may form homodimer, homotetramer and also nuclear bodies and interacts with the transcriptional coactivator CBP and CREBBP. Three isoforms of AIRE are reported and one of them is predicted to be a candidate for nuclear-mediated decay (NMD). AIRE is involved in immune regulation and is known to mediate E3 ubiquitin ligase activity. Defects in AIRE are responsible for the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED).
Specifications

Specifications

AIRE
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O43918
AIRE; autoimmune regulator (automimmune polyendocrinopathy candidiasis ectodermal dystrophy); APS1; APSI; PGA1; APECED; AIRE protein; autoimmune regulator (APECED protein); autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator; AIRE1; OTTHUMP00000109529; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
TM-724
Unconjugated
AIRE
Liquid
AIRE
IgG2a, kappa
25 μg
4° C
Primary
326
Videos
Documents

For Research Use Only.