Learn More
S.C.A.T™ Air Valve for SCAT Safety Caps
The S.C.A.T™ air valve combines valve and filter functions all in one. It blocks harmful vapors and avoids under pressure in your solvent reservoirs. The valve membrane additionally protects your eluents from contamination. The valve is compatible with every generation of SCAT SafetyCaps
Brand:  S.C.A.T™ 117010
Code : NEW
Additional Details : Weight : 0.01880kg
 View more versions of this product
Product Code. 10578823
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Essential element of S.C.A.T™ SafetyCaps
  • Protects from inhalation of harmful vapors
  • Valve membran uncloses for air intake
  • PTFE filter holds back dust and contaminant particles
  • Keeps your mobile phase contamination-free
  • Suitabel for every generation of S.C.A.T™ SafetyCaps
  • Special air valve for preparative HPLC available
  • Fire resistant version available
  • To be changed every 6 months
  • Lab safety, optimal protection when extracting solvents
  • High work performance
  • Technical protective measures
  • Cost efficiency, no need of permanent local extraction
  • Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
  • Technical protective measures
  • Create a comfortable working climate
Specifications
Specifications
150 mL/min
SCAT SafetyCaps
PTFE
Air Valve
HPLC
HDPE, PTFE
1 µ